Anti-RAB6C Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RAB6C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-355
- 77839-354
- Antigen Symbol:
- RAB6C
- RAB6C
- Antigen Name:
- Ras-related protein Rab-6C protein (1-254AA)
- Ras-related protein Rab-6C protein (1-254AA)
- Antigen Synonyms:
- RAB 6C|Rab6 like protein WTH3|Rab6-like protein WTH3|RAB6C|RAB6C member RAS oncogene family|RAB6C_HUMAN|Ras related protein Rab 6C|Ras-related protein Rab-6C|WTH3
- RAB 6C|Rab6 like protein WTH3|Rab6-like protein WTH3|RAB6C|RAB6C member RAS oncogene family|RAB6C_HUMAN|Ras related protein Rab 6C|Ras-related protein Rab-6C|WTH3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H0N0
- Q9H0N0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together