Anti-DDX28 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DDX28
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-776
- 77838-775
- Antigen Symbol:
- DDX28
- DDX28
- Antigen Name:
- Probable ATP-dependent RNA helicase DDX28 protein (71-206AA)
- Probable ATP-dependent RNA helicase DDX28 protein (71-206AA)
- Antigen Synonyms:
- DDX 28|DDX28|DDX28_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 28|DEAD box polypeptide 28|DEAD/H (Asp Glu Ala Asp/His) box polypeptide 28|FLJ11282|MDDX 28|MDDX28|Mitochondrial DEAD box protein 28|Probable ATP-dependent RNA helicase DDX28
- DDX 28|DDX28|DDX28_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 28|DEAD box polypeptide 28|DEAD/H (Asp Glu Ala Asp/His) box polypeptide 28|FLJ11282|MDDX 28|MDDX28|Mitochondrial DEAD box protein 28|Probable ATP-dependent RNA helicase DDX28
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NUL7
- Q9NUL7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together