Anti-KCNK13 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KCNK13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-698
- 77835-697
- Antigen Symbol:
- KCNK13
- KCNK13
- Antigen Name:
- Potassium channel subfamily K member 13 protein (292-408AA)
- Potassium channel subfamily K member 13 protein (292-408AA)
- Antigen Synonyms:
- K2p13.1|K2P13.1 potassium channel|KCNKD|KCNKD_HUMAN|Potassium channel subfamily K member 13|Tandem pore domain halothane inhibited potassium channel 1|Tandem pore domain halothane-inhibited potassium channel 1|Tandem pore domain potassium channel THIK 1|Tandem pore domain potassium channel THIK1|Thik 1|THIK-1|Thik1
- K2p13.1|K2P13.1 potassium channel|KCNKD|KCNKD_HUMAN|Potassium channel subfamily K member 13|Tandem pore domain halothane inhibited potassium channel 1|Tandem pore domain halothane-inhibited potassium channel 1|Tandem pore domain potassium channel THIK 1|Tandem pore domain potassium channel THIK1|Thik 1|THIK-1|Thik1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HB14
- Q9HB14
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together