Anti-Allergen Ara h 1, clone P17 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Allergen Ara h 1, clone P17
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Peanut
Specifications
- Catalog No:
- 77815-896
- 77815-897
- Antigen Symbol:
- Allergen Ara h 1, clone P17
- Allergen Ara h 1, clone P17
- Antigen Name:
- Recombinant Peanut Allergen Ara h 1, clone P17 protein (26-216AA)
- Recombinant Peanut Allergen Ara h 1, clone P17 protein (26-216AA)
- Antigen Synonyms:
- Allergen Ara h 1|clone P17|Allergen Ara h I|allergen Ara h 1
- Allergen Ara h 1|clone P17|Allergen Ara h I|allergen Ara h 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Peanut
- Peanut
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43237
- P43237
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together