Anti-DUSP22 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DUSP22
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77836-473
- 77836-474
- Antigen Symbol:
- DUSP22
- DUSP22
- Antigen Name:
- Dual specificity protein phosphatase 22 protein (1-184AA)
- Dual specificity protein phosphatase 22 protein (1-184AA)
- Antigen Synonyms:
- Dual specificity protein phosphatase 22| DUS22_HUMAN| Dusp22| JNK stimulatory phosphatase 1| JNK-stimulatory phosphatase-1| JSP 1| JSP-1| JSP1| LMW DSP2| LMW-DSP2| Low molecular weight dual specificity phosphatase 2| MAP kinase phosphatase x| Mitogen activated protein kinase phosphatase x| Mitogen-activated protein kinase phosphatase x| MKP-x| MKPX
- Dual specificity protein phosphatase 22| DUS22_HUMAN| Dusp22| JNK stimulatory phosphatase 1| JNK-stimulatory phosphatase-1| JSP 1| JSP-1| JSP1| LMW DSP2| LMW-DSP2| Low molecular weight dual specificity phosphatase 2| MAP kinase phosphatase x| Mitogen activated protein kinase phosphatase x| Mitogen-activated protein kinase phosphatase x| MKP-x| MKPX
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NRW4
- Q9NRW4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together