About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: HSD17B6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77814-635
- 77814-634
- Antigen Symbol:
- HSD17B6
- HSD17B6
- Antigen Name:
- Fusion protein of HSD17B6
- Fusion protein of HSD17B6
- Antigen Synonyms:
- 17-beta-HSD 6|17-beta-HSD6|17-beta-hydroxysteroid dehydrogenase type 6|3 hydroxysteroid epimerase|3(alpha >beta) hydroxysteroid epimerase|3(alpha >beta) hydroxysteroid epimerasel|3-alpha->,beta-HSE|3-alpha->,beta-hydroxysteroid epimerase|H17B6_HUMAN|HSD17B6|HSE|Hydroxysteroid (17 beta) dehydrogenase 6|Hydroxysteroid (17 beta) dehydrogenase 6 homolog (mouse)|Hydroxysteroid 17 beta dehydrogenase 6|NAD+ dependent 3 alpha hydroxysteroid dehydrogenase 3 hydroxysteroid epimerase|NAD+ dependent 3 alpha hydroxysteroid dehydrogenase|Oxidative 3 alpha hydroxysteroid dehydrogenase|Oxidative 3-alpha hydroxysteroid dehydrogenase|Oxidoreductase|Retinol dehydrogenase|RODH|SDR9C6|Short chain dehydrogenase/reductase family 9C, member 6
- 17-beta-HSD 6|17-beta-HSD6|17-beta-hydroxysteroid dehydrogenase type 6|3 hydroxysteroid epimerase|3(alpha >beta) hydroxysteroid epimerase|3(alpha >beta) hydroxysteroid epimerasel|3-alpha->,beta-HSE|3-alpha->,beta-hydroxysteroid epimerase|H17B6_HUMAN|HSD17B6|HSE|Hydroxysteroid (17 beta) dehydrogenase 6|Hydroxysteroid (17 beta) dehydrogenase 6 homolog (mouse)|Hydroxysteroid 17 beta dehydrogenase 6|NAD+ dependent 3 alpha hydroxysteroid dehydrogenase 3 hydroxysteroid epimerase|NAD+ dependent 3 alpha hydroxysteroid dehydrogenase|Oxidative 3 alpha hydroxysteroid dehydrogenase|Oxidative 3-alpha hydroxysteroid dehydrogenase|Oxidoreductase|Retinol dehydrogenase|RODH|SDR9C6|Short chain dehydrogenase/reductase family 9C, member 6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14756
- O14756
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice