Anti-AATF Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: AATF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77808-531
- 77808-532
- Antigen Symbol:
- AATF
- AATF
- Antigen Name:
- Protein AATF protein (6-294AA)
- Protein AATF protein (6-294AA)
- Antigen Synonyms:
- AATF|AATF_HUMAN|Apoptosis antagonizing transcription factor|Apoptosis-antagonizing transcription factor|BFR2|CHE 1|CHE1|DED|Protein AATF|Rb binding protein Che 1|Rb-binding protein Che-1
- AATF|AATF_HUMAN|Apoptosis antagonizing transcription factor|Apoptosis-antagonizing transcription factor|BFR2|CHE 1|CHE1|DED|Protein AATF|Rb binding protein Che 1|Rb-binding protein Che-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NY61
- Q9NY61
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together