Anti-ORM1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: ORM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77808-849
- 77808-848
- Antigen Symbol:
- ORM1
- ORM1
- Antigen Name:
- Alpha-1-acid glycoprotein 1 protein (19-201AA)
- Alpha-1-acid glycoprotein 1 protein (19-201AA)
- Antigen Synonyms:
- A1AG1_HUMAN| AGP 1| AGP A| AGP| AGP1| Alpha 1 acid glycoprotein| Alpha-1-acid glycoprotein 1| alpha-1-AGP| Epididymis secretory sperm binding protein Li 153w| glycoprotein| alpha-1-acid| of serum| HEL S 153w| OMD 1| ORM| ORM1| Orosomucoid 1| Orosomucoid-1
- A1AG1_HUMAN| AGP 1| AGP A| AGP| AGP1| Alpha 1 acid glycoprotein| Alpha-1-acid glycoprotein 1| alpha-1-AGP| Epididymis secretory sperm binding protein Li 153w| glycoprotein| alpha-1-acid| of serum| HEL S 153w| OMD 1| ORM| ORM1| Orosomucoid 1| Orosomucoid-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02763
- P02763
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together