Anti-RUNX1T1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: RUNX1T1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77809-936
- 77809-935
- Antigen Symbol:
- RUNX1T1
- RUNX1T1
- Antigen Name:
- Synthetic peptide of RUNX1T1
- Synthetic peptide of RUNX1T1
- Antigen Synonyms:
- Acute myelogenous leukemia 1 translocation 1 cyclin D related|Acute myelogenous leukemia 1 translocation 1 protein|AML1 MTG8|AML1T1|CBFA2T1|CDR|Core binding factor runt domain alpha subunit 2 translocated to 1|Core binding factor runt domain alpha subunit 2 translocated to 1 cyclin D related|Cyclin D related|Cyclin D related protein|Cyclin-D-related protein|Eight twenty one protein|ETO|ETO protein|MGC2796|MTG 8|MTG 8b|MTG8|MTG8 protein|MTG8_HUMAN|MTG8b|Myeloid translocation gene on 8q22|Protein CBFA2T1|Protein ETO|Protein MTG 8|Protein MTG8|Runt related transcription factor 1 translocated to 1 (cyclin D related)|Runt related transcription factor 1 translocated to 1|Runt related transcription factor 1 translocated to 1 cyclin D related|RUNX1 translocation partner 1|RUNX1T1|Zinc finger MYND domain containing protein 2|Zinc finger MYND domain-containing protein 2|ZMYND 2|ZMYND2
- Acute myelogenous leukemia 1 translocation 1 cyclin D related|Acute myelogenous leukemia 1 translocation 1 protein|AML1 MTG8|AML1T1|CBFA2T1|CDR|Core binding factor runt domain alpha subunit 2 translocated to 1|Core binding factor runt domain alpha subunit 2 translocated to 1 cyclin D related|Cyclin D related|Cyclin D related protein|Cyclin-D-related protein|Eight twenty one protein|ETO|ETO protein|MGC2796|MTG 8|MTG 8b|MTG8|MTG8 protein|MTG8_HUMAN|MTG8b|Myeloid translocation gene on 8q22|Protein CBFA2T1|Protein ETO|Protein MTG 8|Protein MTG8|Runt related transcription factor 1 translocated to 1 (cyclin D related)|Runt related transcription factor 1 translocated to 1|Runt related transcription factor 1 translocated to 1 cyclin D related|RUNX1 translocation partner 1|RUNX1T1|Zinc finger MYND domain containing protein 2|Zinc finger MYND domain-containing protein 2|ZMYND 2|ZMYND2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q06455
- Q06455
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together