Anti-TEAD4 Rabbit Polyclonal Antibody
Catalog # 77802-195
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Transcriptional enhancer factor TEF3
- Antigen Symbol:TEAD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q15561
- Antigen Synonyms:EFTR 2|EFTR2|hRTEF 1B|hRTEF1B|MGC9014|OTTHUMP00000238119|OTTHUMP00000238122|OTTHUMP00000238124|Related to TEF 1|Related to TEF1|Related transcription enhancer factor 1B|RTEF1|TCF13L1|TEA domain family member 4|TEAD 4|TEAD-4|TEAD4|TEAD4_HUMAN|TEF 3|TEF3|TEFR 1|TEFR1|Transcription factor 13 (SV40 transcriptional enhancer factor) like 1|Transcription factor 13 like 1|Transcription factor 13-like 1|Transcription factor RTEF 1|Transcription factor RTEF-1|Transcription factor RTEF1|Transcriptional enhancer factor 1 related|Transcriptional enhancer factor 3|Transcriptional enhancer factor TEF 3|Transcriptional enhancer factor TEF-3|Transcriptional enhancer factor TEF3
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TEAD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat