Anti-MLST8 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MLST8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-882
- 77835-881
- Antigen Symbol:
- MLST8
- MLST8
- Antigen Name:
- Target of rapamycin complex subunit LST8 protein (159-220AA)
- Target of rapamycin complex subunit LST8 protein (159-220AA)
- Antigen Synonyms:
- G protein beta subunit like| G protein beta subunit-like| Gable| GbetaL| GBL| GBL protein| LST8| LST8_HUMAN| Mammalian lethal with SEC13 protein 8| MGC111011| mLST8| MTOR associated protein LST8 homolog (S. cerevisiae)| POP3| POP3 homolog (S. pombe)| Protein GbetaL| Target of rapamycin complex subunit LST8| TORC subunit LST8| WAT1| WAT1 homolog (S. pombe)
- G protein beta subunit like| G protein beta subunit-like| Gable| GbetaL| GBL| GBL protein| LST8| LST8_HUMAN| Mammalian lethal with SEC13 protein 8| MGC111011| mLST8| MTOR associated protein LST8 homolog (S. cerevisiae)| POP3| POP3 homolog (S. pombe)| Protein GbetaL| Target of rapamycin complex subunit LST8| TORC subunit LST8| WAT1| WAT1 homolog (S. pombe)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BVC4
- Q9BVC4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together