Anti-HSD17B10 Rabbit Polyclonal Antibody
Catalog # 77827-225
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:ERAB
- Antigen Symbol:HSD17B10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q99714
- Antigen Synonyms:17 beta hydroxysteroid dehydrogenase 10|17 beta hydroxysteroid dehydrogenase type 10|17-beta-HSD 10|17-beta-hydroxysteroid dehydrogenase 10|17b HSD10|3 hydroxy 2 methylbutyryl CoA dehydrogenase|3 hydroxyacyl CoA dehydrogenase type 2|3 hydroxyacyl CoA dehydrogenase type II|3-hydroxy-2-methylbutyryl-CoA dehydrogenase|3-hydroxyacyl-CoA dehydrogenase type II|3-hydroxyacyl-CoA dehydrogenase type-2|AB binding alcohol dehydrogenase|ABAD|Ads9|Amyloid beta binding polypeptide|Amyloid beta peptide binding alcohol dehydrogenase|Amyloid beta peptide binding protein|CAMR|DUPXp11.22|Endoplasmic Reticulum Amyloid Binding Protein|Endoplasmic reticulum associated amyloid beta peptide binding protein|Endoplasmic reticulum-associated amyloid beta-peptide-binding protein|ER associated amyloid beta-binding protein|ERAB|HADH 2|HADH2|HCD 2|HCD2|HCD2_HUMAN|Hsd17b10|Hydroxyacyl CoA Dehydrogenase type II|Hydroxyacyl Coenzyme A dehydrogenase type II|Hydroxysteroid (17 beta) dehydrogenase 10|Mental retardation X linked syndromic 11|MHBD|Mitochondrial L3 Hydroxyacyl CoA Dehydrogenase|Mitochondrial ribonuclease P protein 2|Mitochondrial RNase P protein 2|MRPP2|MRX17|SCHAD|SDR5C1|Short chain dehydrogenase/reductase family 5C member 1|Short chain L 3 hydroxyacyl CoA dehydrogenase type 2|Short chain type dehydrogenase/reductase XH98G2|Short-chain type dehydrogenase/reductase XH98G2|Type 10 17b HSD|Type 10 17beta hydroxysteroid dehydrogenase|Type II HADH|XH98G2
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:1000
Type:
Antigen: HSD17B10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat