Anti-HTRA3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:5000, IHC:1:50 to 1:200
Type:
Antigen: HTRA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77821-338
- 77821-337
- Antigen Symbol:
- HTRA3
- HTRA3
- Antigen Name:
- Synthetic peptide of HTRA3
- Synthetic peptide of HTRA3
- Antigen Synonyms:
- High temperature requirement factor A3|High-temperature requirement factor A3|HtrA serine peptidase 3|HTRA3|HTRA3_HUMAN|Pregnancy related serine protease|Pregnancy-related serine protease|Probable serine protease HTRA3|PRSP|Serine protease HTRA3|Tasp
- High temperature requirement factor A3|High-temperature requirement factor A3|HtrA serine peptidase 3|HTRA3|HTRA3_HUMAN|Pregnancy related serine protease|Pregnancy-related serine protease|Probable serine protease HTRA3|PRSP|Serine protease HTRA3|Tasp
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P83110
- P83110
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together