Anti-PKN3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PKN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-718
- 77829-717
- Antigen Symbol:
- PKN3
- PKN3
- Antigen Name:
- Serine/threonine-protein kinase N3 protein (1-252AA)
- Serine/threonine-protein kinase N3 protein (1-252AA)
- Antigen Synonyms:
- pkn3| PKN3_HUMAN| PKNBETA| Protein kinase C related kinase 3| Protein kinase N3| Protein kinase PKN beta| Protein kinase PKN-beta| Protein-kinase C-related kinase 3| RP11-545E17.1| Serine/threonine protein kinase N3| Serine/threonine-protein kinase N3
- pkn3| PKN3_HUMAN| PKNBETA| Protein kinase C related kinase 3| Protein kinase N3| Protein kinase PKN beta| Protein kinase PKN-beta| Protein-kinase C-related kinase 3| RP11-545E17.1| Serine/threonine protein kinase N3| Serine/threonine-protein kinase N3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P5Z2
- Q6P5Z2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together