Anti-porB Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: porB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Neisseria meningitidis serogroup B, serotype 15
Specifications
- Catalog No:
- 77819-153
- 77819-154
- Antigen Symbol:
- porB
- porB
- Antigen Name:
- Recombinant Neisseria meningitidis serogroup B / serotype 15 Major outer membrane protein P.IB protein (20-331AA)
- Recombinant Neisseria meningitidis serogroup B / serotype 15 Major outer membrane protein P.IB protein (20-331AA)
- Antigen Synonyms:
- porB| NMBH4476_1982| NMH_2035| Major outer membrane protein P.IB| PIB| Protein IB| Class 3 protein| Porin
- porB| NMBH4476_1982| NMH_2035| Major outer membrane protein P.IB| PIB| Protein IB| Class 3 protein| Porin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Neisseria meningitidis serogroup B, serotype 15
- Neisseria meningitidis serogroup B, serotype 15
- Host:
- Rabbit
- Rabbit
- Gene ID:
- E6MZM0
- E6MZM0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together