Anti-ST6GALNAC5 Rabbit Polyclonal Antibody
77801-186
New Product
- Antigen Name:ST6GalNAcV
- Antigen Symbol:ST6GALNAC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BVH7
- Antigen Synonyms:6-sialyltransferase 5|6-sialyltransferase V|Alpha N acetylgalactosaminide alpha 2,6 sialyltransferase 5|Alpha-N-acetylgalactosaminide alpha-2|GalNAc alpha 2,6 sialyltransferase V|GalNAc alpha-2|GD1 alpha synthase|SIA7E_HUMAN|Sialyltransferase 7E|Sialyltransferase E|SIAT7 E|SIAT7-E|SIAT7E|ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5|ST6GalNAc V|St6galnac5|ST6GalNAcV
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: ST6GALNAC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat