Anti-SLC6A15 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC6A15
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-407
- 77839-406
- Antigen Symbol:
- SLC6A15
- SLC6A15
- Antigen Name:
- Sodium-dependent neutral amino acid transporter B(0)AT2 protein (357-452AA)
- Sodium-dependent neutral amino acid transporter B(0)AT2 protein (357-452AA)
- Antigen Synonyms:
- hv7 3|MGC87066|NTT73|Orphan sodium and chloride dependent neurotransmitter transporter NTT73|Orphan transporter v7 3|S6A15_HUMAN|SBAT1|SLC6A15|sodium- and chloride-dependent neurotransmitter transporter NTT73|Sodium-coupled branched-chain amino-acid transporter 1|Sodium-dependent neutral amino acid transporter B(0)AT2|Solute carrier family 6 (neutral amino acid transporter) member 15|Solute carrier family 6 member 15|Transporter v7-3|V7 3
- hv7 3|MGC87066|NTT73|Orphan sodium and chloride dependent neurotransmitter transporter NTT73|Orphan transporter v7 3|S6A15_HUMAN|SBAT1|SLC6A15|sodium- and chloride-dependent neurotransmitter transporter NTT73|Sodium-coupled branched-chain amino-acid transporter 1|Sodium-dependent neutral amino acid transporter B(0)AT2|Solute carrier family 6 (neutral amino acid transporter) member 15|Solute carrier family 6 member 15|Transporter v7-3|V7 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2J7
- Q9H2J7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together