Anti-POU2AF1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POU2AF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-180
- 77824-179
- Antigen Symbol:
- POU2AF1
- POU2AF1
- Antigen Name:
- POU domain class 2-associating factor 1 protein (1-256AA)
- POU domain class 2-associating factor 1 protein (1-256AA)
- Antigen Synonyms:
- B cell Oct binding protein 1|B cell specific coactivator OBF 1|B cell specific coactivator OBF1|B-cell-specific coactivator OBF-1|BOB 1|BOB-1|OBF 1|OBF1|OBF1_HUMAN|OCA B|OCA-B|OCAB|OCT binding factor 1|OCT-binding factor 1|POU class 2 associating factor 1|POU domain class 2 associating factor 1|POU domain class 2-associating factor 1|Pou2af1
- B cell Oct binding protein 1|B cell specific coactivator OBF 1|B cell specific coactivator OBF1|B-cell-specific coactivator OBF-1|BOB 1|BOB-1|OBF 1|OBF1|OBF1_HUMAN|OCA B|OCA-B|OCAB|OCT binding factor 1|OCT-binding factor 1|POU class 2 associating factor 1|POU domain class 2 associating factor 1|POU domain class 2-associating factor 1|Pou2af1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16633
- Q16633
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together