Anti-KRT81 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:15 to 1:50
Type:
Antigen: KRT81
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77807-636
- 77807-637
- Antigen Symbol:
- KRT81
- KRT81
- Antigen Name:
- Fusion protein of KRT81
- Fusion protein of KRT81
- Antigen Synonyms:
- 1|basic|ghHb 1|ghHb1|ghHkb 1|ghHKb1|hair|Hair keratin K2.9|Hard keratin type II 1|HB 1|HB1|hHAKB2 1|K2.9|K81|Keratin 81|Keratin|Keratin hair basic 1|Keratin type II cuticular Hb1|Keratin-81|Keratin81|KRT 81|KRT81|KRT81_HUMAN|KRTHB 1|KRTHB1|Metastatic lymph node 137 gene protein|MLN 137|MLN137|type II cuticular Hb1|Type II hair keratin Hb1|Type-II keratin Kb21
- 1|basic|ghHb 1|ghHb1|ghHkb 1|ghHKb1|hair|Hair keratin K2.9|Hard keratin type II 1|HB 1|HB1|hHAKB2 1|K2.9|K81|Keratin 81|Keratin|Keratin hair basic 1|Keratin type II cuticular Hb1|Keratin-81|Keratin81|KRT 81|KRT81|KRT81_HUMAN|KRTHB 1|KRTHB1|Metastatic lymph node 137 gene protein|MLN 137|MLN137|type II cuticular Hb1|Type II hair keratin Hb1|Type-II keratin Kb21
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14533
- Q14533
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together