Anti-SRFBP1 Rabbit Polyclonal Antibody
Catalog # 77800-971
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:p49 / STRAP
- Antigen Symbol:SRFBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:Q8NEF9
- Antigen Synonyms:p49 / STRAP|p49|p49/STRAP|serum response factor binding protein 1|Serum response factor-binding protein 1|SRF dependent transcription regulation associated protein|SRF-dependent transcription regulation-associated protein|SRFB1_HUMAN|SRFBP 1|SRFBP1
- Storage Buffer:PBS, 0.1% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SRFBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human