Order Entry
United States
ContactUsLinkComponent
Anti-ZNRD1 Rabbit Recombinant Polyclonal Antibody
Anti-ZNRD1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-ZNRD1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77842-579
  • 77842-580
  • Antigen Symbol:
  • ZNRD1
  • ZNRD1
  • Antigen Name:
  • DNA-directed RNA polymerase I subunit RPA12 protein (1-126AA)
  • DNA-directed RNA polymerase I subunit RPA12 protein (1-126AA)
  • Antigen Synonyms:
  • DNA directed RNA polymerase I subunit RPA12|DNA-directed RNA polymerase I subunit RPA12|HTEX 6|hZR14|RNA polymerase I small specific subunit Rpa12|RPA12|RPA12_HUMAN|tctex 6|TCTEX6|TEX6|Transcription associated zinc ribbon protein|Zinc ribbon domain containing 1|Zinc ribbon domain containing protein 1|Zinc ribbon domain-containing protein 1|ZNRD1|ZR14
  • DNA directed RNA polymerase I subunit RPA12|DNA-directed RNA polymerase I subunit RPA12|HTEX 6|hZR14|RNA polymerase I small specific subunit Rpa12|RPA12|RPA12_HUMAN|tctex 6|TCTEX6|TEX6|Transcription associated zinc ribbon protein|Zinc ribbon domain containing 1|Zinc ribbon domain containing protein 1|Zinc ribbon domain-containing protein 1|ZNRD1|ZR14
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9P1U0
  • Q9P1U0
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Storage Buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together