About this item
Recommended dilution: ELISA:1:1000 to 1:5000, WB:1:200 to 1:1000, IHC:1:10 to 1:50
Type:
Antigen: MSH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77811-568
- 77803-568
- Antigen Symbol:
- MSH3
- MSH3
- Antigen Name:
- Fusion protein of MSH3
- Fusion protein of MSH3
- Antigen Synonyms:
- Divergent upstream protein|DNA mismatch repair protein|DNA mismatch repair protein Msh 3|DNA mismatch repair protein MSH3|DUC 1|DUC1|DUG|DUP|hMSH3|MGC163306|MGC163308|Mismatch repair protein 1|MRP 1|MRP1|MSH 3|MSH3|MSH3_HUMAN|MutS homolog 3 (E. coli)|MutS homolog 3
- Divergent upstream protein|DNA mismatch repair protein|DNA mismatch repair protein Msh 3|DNA mismatch repair protein MSH3|DUC 1|DUC1|DUG|DUP|hMSH3|MGC163306|MGC163308|Mismatch repair protein 1|MRP 1|MRP1|MSH 3|MSH3|MSH3_HUMAN|MutS homolog 3 (E. coli)|MutS homolog 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20585
- P20585
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice