Anti-PNCK Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PNCK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-619
- 77830-618
- Antigen Symbol:
- PNCK
- PNCK
- Antigen Name:
- Calcium/calmodulin-dependent protein kinase type 1B protein (1-115AA)
- Calcium/calmodulin-dependent protein kinase type 1B protein (1-115AA)
- Antigen Synonyms:
- Calcium/calmodulin dependent protein kinase type 1B| Calcium/calmodulin-dependent protein kinase type 1B| CaM KI beta| CaM kinase I beta| CaM kinase IB| CaM-KI beta| CaMK1b| CaMKI beta| CaMKI-beta| EC 2.7.11.17| KCC1B_HUMAN| MGC45419| PNCK| Pregnancy up regulated non ubiquitously expressed CaM kinase| Pregnancy up-regulated non-ubiquitously-expressed CaM kinase
- Calcium/calmodulin dependent protein kinase type 1B| Calcium/calmodulin-dependent protein kinase type 1B| CaM KI beta| CaM kinase I beta| CaM kinase IB| CaM-KI beta| CaMK1b| CaMKI beta| CaMKI-beta| EC 2.7.11.17| KCC1B_HUMAN| MGC45419| PNCK| Pregnancy up regulated non ubiquitously expressed CaM kinase| Pregnancy up-regulated non-ubiquitously-expressed CaM kinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P2M8
- Q6P2M8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together