Anti-UGT1A1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UGT1A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-960
- 77803-960
- Antigen Symbol:
- UGT1A1
- UGT1A1
- Antigen Name:
- UDP-glucuronosyltransferase 1-1 protein (395-482AA)
- UDP-glucuronosyltransferase 1-1 protein (395-482AA)
- Antigen Synonyms:
- BILIQTL1|Bilirubin specific UDPGT isozyme 1|bilirubin UDP glucuronosyltransferase 1 1|bilirubin UDP glucuronosyltransferase isozyme 1|Bilirubin-specific UDPGT isozyme 1|EC 2.4.1.17|GNT1|HUG BR1|HUG-BR1|HUGBR1|PHENOL/BILIRUBIN UDP GLUCURONOSYLTRANSFERASE|UD11_HUMAN|UDP glucuronosyltransferase 1 1 [Precursor]|UDP glucuronosyltransferase 1 family polypeptide A1|UDP glucuronosyltransferase 1A1|UDP GLYCOSYLTRANSFERASE 1|UDP-glucuronosyltransferase 1-1|UDP-glucuronosyltransferase 1-A|UDP-glucuronosyltransferase 1A1|UDPGT|UDPGT 1-1|UGT 1A|UGT-1A|UGT1|UGT1 01|UGT1*1|UGT1-01|UGT1.1|UGT1A|Ugt1a1|URIDINE DIPHOSPHATE GLUCURONOSYLTRANSFERASE, BILIRUBIN/PHENOL|URIDINE DIPHOSPHATE GLYCOSYLTRANSFERASE 1|URIDINE DIPHOSPHATE GLYCOSYLTRANSFERASE 1 FAMILY, POLYPEPTIDE A1
- BILIQTL1|Bilirubin specific UDPGT isozyme 1|bilirubin UDP glucuronosyltransferase 1 1|bilirubin UDP glucuronosyltransferase isozyme 1|Bilirubin-specific UDPGT isozyme 1|EC 2.4.1.17|GNT1|HUG BR1|HUG-BR1|HUGBR1|PHENOL/BILIRUBIN UDP GLUCURONOSYLTRANSFERASE|UD11_HUMAN|UDP glucuronosyltransferase 1 1 [Precursor]|UDP glucuronosyltransferase 1 family polypeptide A1|UDP glucuronosyltransferase 1A1|UDP GLYCOSYLTRANSFERASE 1|UDP-glucuronosyltransferase 1-1|UDP-glucuronosyltransferase 1-A|UDP-glucuronosyltransferase 1A1|UDPGT|UDPGT 1-1|UGT 1A|UGT-1A|UGT1|UGT1 01|UGT1*1|UGT1-01|UGT1.1|UGT1A|Ugt1a1|URIDINE DIPHOSPHATE GLUCURONOSYLTRANSFERASE, BILIRUBIN/PHENOL|URIDINE DIPHOSPHATE GLYCOSYLTRANSFERASE 1|URIDINE DIPHOSPHATE GLYCOSYLTRANSFERASE 1 FAMILY, POLYPEPTIDE A1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P22309
- P22309
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together