Anti-IL12A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:10 to 1:50
Type:
Antigen: IL12A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77839-659
- 77839-658
- Antigen Symbol:
- IL12A
- IL12A
- Antigen Name:
- Synthetic peptide of IL12A
- Synthetic peptide of IL12A
- Antigen Synonyms:
- CLMF|CLMF p35|CLMF p40|CLMF1|CLMF2|Cytotoxic lymphocyte maturation factor 1, p35|Cytotoxic lymphocyte maturation factor 2|Cytotoxic lymphocyte maturation factor 35 kDa subunit|Cytotoxic lymphocyte maturation factor 40 kDa subunit|IL 12A|IL 2 subunit p40|IL-12 subunit p35|IL-12A|IL-12B|IL12|IL12, p35 subunit|IL12, subunit p40|IL12A|IL12A_HUMAN|IL12B|IL35 subunit|Interleukin 12 alpha chain|Interleukin 12 beta chain|Interleukin 12 subunit beta|Interleukin 12, p40|Interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35)|Interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40)|Interleukin 12B|Interleukin-12 subunit alpha|Natural killer cell stimulatory factor 1, 35 kD subunit|natural killer cell stimulatory factor 2|Natural killer cell stimulatory factor, 40 kD subunit|NF cell stimulatory factor chain 1|NFSK|NK cell stimulatory factor chain 1|NK cell stimulatory factor chain 2|NKSF|NKSF1|NKSF2|P35
- CLMF|CLMF p35|CLMF p40|CLMF1|CLMF2|Cytotoxic lymphocyte maturation factor 1, p35|Cytotoxic lymphocyte maturation factor 2|Cytotoxic lymphocyte maturation factor 35 kDa subunit|Cytotoxic lymphocyte maturation factor 40 kDa subunit|IL 12A|IL 2 subunit p40|IL-12 subunit p35|IL-12A|IL-12B|IL12|IL12, p35 subunit|IL12, subunit p40|IL12A|IL12A_HUMAN|IL12B|IL35 subunit|Interleukin 12 alpha chain|Interleukin 12 beta chain|Interleukin 12 subunit beta|Interleukin 12, p40|Interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35)|Interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40)|Interleukin 12B|Interleukin-12 subunit alpha|Natural killer cell stimulatory factor 1, 35 kD subunit|natural killer cell stimulatory factor 2|Natural killer cell stimulatory factor, 40 kD subunit|NF cell stimulatory factor chain 1|NFSK|NK cell stimulatory factor chain 1|NK cell stimulatory factor chain 2|NKSF|NKSF1|NKSF2|P35
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P29459
- P29459
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together