Anti-RAB27B Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAB27B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-366
- 77796-365
- Antigen Symbol:
- RAB27B
- RAB27B
- Antigen Name:
- Ras-related protein Rab-27B protein (2-218AA)
- Ras-related protein Rab-27B protein (2-218AA)
- Antigen Synonyms:
- C25KG|Rab27b|RAB27B member RAS oncogene family|RAS associated protein RAB27B|Ras related protein Rab 27b|Ras related protein Rab27b|Ras-related protein Rab-27B|RB27B_HUMAN|Small GTP binding protein rab27b
- C25KG|Rab27b|RAB27B member RAS oncogene family|RAS associated protein RAB27B|Ras related protein Rab 27b|Ras related protein Rab27b|Ras-related protein Rab-27B|RB27B_HUMAN|Small GTP binding protein rab27b
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00194
- O00194
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together