Anti-PTPRZ1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PTPRZ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-179
- 77796-180
- Antigen Symbol:
- PTPRZ1
- PTPRZ1
- Antigen Name:
- Receptor-type tyrosine-protein phosphatase zeta protein (36-300AA)
- Receptor-type tyrosine-protein phosphatase zeta protein (36-300AA)
- Antigen Synonyms:
- 3F8 chondroitin sulfate proteoglycan|3H1 keratan sulfate proteoglycan|HPTPZ|HPTPzeta|Phosphacan|Protein tyrosine phosphatase receptor type Z polypeptide 2|Protein tyrosine phosphatase, receptor type, Z polypeptide 1|Protein tyrosine phosphatase, receptor type, zeta polypeptide 1|Protein-tyrosine phosphatase receptor type Z polypeptide 1|Protein-tyrosine phosphatase receptor type Z polypeptide 2|PTP-ZETA|PTP18|PTPRZ|PTPRZ_HUMAN|Ptprz1|PTPZ|R PTP zeta 2|R-PTP-zeta|R-PTP-zeta-2|Receptor type tyrosine phosphatase beta/zeta|Receptor-type tyrosine-protein phosphatase zeta|RPTP-BETA|RPTPB|RPTPbeta
- 3F8 chondroitin sulfate proteoglycan|3H1 keratan sulfate proteoglycan|HPTPZ|HPTPzeta|Phosphacan|Protein tyrosine phosphatase receptor type Z polypeptide 2|Protein tyrosine phosphatase, receptor type, Z polypeptide 1|Protein tyrosine phosphatase, receptor type, zeta polypeptide 1|Protein-tyrosine phosphatase receptor type Z polypeptide 1|Protein-tyrosine phosphatase receptor type Z polypeptide 2|PTP-ZETA|PTP18|PTPRZ|PTPRZ_HUMAN|Ptprz1|PTPZ|R PTP zeta 2|R-PTP-zeta|R-PTP-zeta-2|Receptor type tyrosine phosphatase beta/zeta|Receptor-type tyrosine-protein phosphatase zeta|RPTP-BETA|RPTPB|RPTPbeta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P23471
- P23471
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together