Anti-LZTFL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LZTFL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-287
- 77842-286
- Antigen Symbol:
- LZTFL1
- LZTFL1
- Antigen Name:
- Leucine zipper transcription factor-like protein 1 protein (116-299AA)
- Leucine zipper transcription factor-like protein 1 protein (116-299AA)
- Antigen Synonyms:
- 5530402H04Rik|6130400H19Rik|AI414725|AW048545|BBS17|DKFZp686H2076|FLJ36386|Leucine zipper transcription factor like 1|Leucine zipper transcription factor like protein 1|Leucine zipper transcription factor-like protein 1|LZTFL 1|Lztfl1|Lztfl1 leucine zipper transcription factor-like 1|LZTL1_HUMAN|MGC106871|MGC108960|Putative uncharacterized protein DKFZp686H2076
- 5530402H04Rik|6130400H19Rik|AI414725|AW048545|BBS17|DKFZp686H2076|FLJ36386|Leucine zipper transcription factor like 1|Leucine zipper transcription factor like protein 1|Leucine zipper transcription factor-like protein 1|LZTFL 1|Lztfl1|Lztfl1 leucine zipper transcription factor-like 1|LZTL1_HUMAN|MGC106871|MGC108960|Putative uncharacterized protein DKFZp686H2076
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQ48
- Q9NQ48
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together