Anti-VLDLR Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VLDLR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-386
- 77804-385
- Antigen Symbol:
- VLDLR
- VLDLR
- Antigen Name:
- Very low-density lipoprotein receptor protein (133-247AA)
- Very low-density lipoprotein receptor protein (133-247AA)
- Antigen Synonyms:
- FLJ35024|Very low density lipoprotein receptor|Very low-density lipoprotein receptor|VLDL R|VLDL receptor|VLDL-R|VLDLR|VLDLR_HUMAN|VLDLRCH
- FLJ35024|Very low density lipoprotein receptor|Very low-density lipoprotein receptor|VLDL R|VLDL receptor|VLDL-R|VLDLR|VLDLR_HUMAN|VLDLRCH
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P98155
- P98155
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together