Anti-MMP23A Rabbit Polyclonal Antibody
77810-313
New Product
- Antigen Name:Synthesized peptide derived from the C-terminal region of MMP-23
- Antigen Symbol:MMP23A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O75900
- Antigen Synonyms:Femalysin|Matrix metallopeptidase 21|matrix metallopeptidase 23B|matrix metalloproteinase 22|formerly|Matrix metalloproteinase 23|soluble form|Matrix metalloproteinase 23B|matrix metalloproteinase in the female reproductive tract|Matrix metalloproteinase-21|Matrix metalloproteinase-22|Matrix metalloproteinase-23|MIFR 1|MIFR|MIFR-1|MMP 21|MMP 22|MMP 23|MMP-21|MMP-22|MMP-23|MMP21|MMP22|MMP23|MMP23_HUMAN|MMP23A|MMP23B
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: MMP23A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat