Anti-NDUFA12 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200
Type:
Antigen: NDUFA12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77841-448
- 77841-447
- Antigen Symbol:
- NDUFA12
- NDUFA12
- Antigen Name:
- NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12 protein (1-145AA)
- NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12 protein (1-145AA)
- Antigen Synonyms:
- 13 kDa differentiation associated protein|13 kDa differentiation-associated protein|2410011G03Rik|AW112974|B17.2|CI-B17.2|CIB17.2|Complex I B17.2|Complex I B17.2 subunit|Complex I-B17.2|DAP13|MGC107642|MGC7999|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 12|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12|NADH ubiquinone oxidoreductase subunit B17.2|NADH-ubiquinone oxidoreductase subunit B17.2|NDUAC_HUMAN|NDUFA12|RGD1311462
- 13 kDa differentiation associated protein|13 kDa differentiation-associated protein|2410011G03Rik|AW112974|B17.2|CI-B17.2|CIB17.2|Complex I B17.2|Complex I B17.2 subunit|Complex I-B17.2|DAP13|MGC107642|MGC7999|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 12|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12|NADH ubiquinone oxidoreductase subunit B17.2|NADH-ubiquinone oxidoreductase subunit B17.2|NDUAC_HUMAN|NDUFA12|RGD1311462
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UI09
- Q9UI09
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together