Anti-PRSS23 Rabbit Recombinant Polyclonal Antibody
Catalog # 77795-649
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Serine protease 23 protein (20-383AA)
- Antigen Symbol:PRSS23
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O95084
- Antigen Synonyms:Protease serine 23|PRS23_HUMAN|Prss23|Putative secreted protein ZSIG13|Serine protease 23|Serine protease umbilical endothelium|SIG13|SPUVE|UNQ270/PRO307|ZSIG13
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PRSS23
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human