Anti-MMP19 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: MMP19
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77834-410
- 77834-409
- Antigen Symbol:
- MMP19
- MMP19
- Antigen Name:
- Matrix metalloproteinase-19 protein (209-508AA)
- Matrix metalloproteinase-19 protein (209-508AA)
- Antigen Synonyms:
- Matrix metalloproteinase 18| Matrix metalloproteinase 19 isoform rasi1| Matrix metalloproteinase 19 isoform rasi9| Matrix metalloproteinase RASI| Matrix metalloproteinase-18| Matrix metalloproteinase-19| MMP-18| MMP-19| MMP18| MMP19| MMP19_HUMAN| Preproprotein matrix metalloproteinase 19 isoform rasi6| RASI 1| RASI1
- Matrix metalloproteinase 18| Matrix metalloproteinase 19 isoform rasi1| Matrix metalloproteinase 19 isoform rasi9| Matrix metalloproteinase RASI| Matrix metalloproteinase-18| Matrix metalloproteinase-19| MMP-18| MMP-19| MMP18| MMP19| MMP19_HUMAN| Preproprotein matrix metalloproteinase 19 isoform rasi6| RASI 1| RASI1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99542
- Q99542
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together