Anti-SCO1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SCO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-853
- 77798-852
- Antigen Symbol:
- SCO1
- SCO1
- Antigen Name:
- Protein SCO1 homolog, mitochondrial protein (29-176AA)
- Protein SCO1 homolog, mitochondrial protein (29-176AA)
- Antigen Synonyms:
- Cytochrome oxidase deficient homolog|Cytochrome oxidase deficient homolog 1|Protein SCO1 homolog mitochondrial|Protein SCO1 homolog, mitochondrial|SCO (cytochrome oxidase deficient yeast) homolog 1|SCO cytochrome oxidase deficient homolog 1 (yeast)|SCO cytochrome oxidase deficient homolog 1|sco1|SCO1_HUMAN|SCOD1
- Cytochrome oxidase deficient homolog|Cytochrome oxidase deficient homolog 1|Protein SCO1 homolog mitochondrial|Protein SCO1 homolog, mitochondrial|SCO (cytochrome oxidase deficient yeast) homolog 1|SCO cytochrome oxidase deficient homolog 1 (yeast)|SCO cytochrome oxidase deficient homolog 1|sco1|SCO1_HUMAN|SCOD1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75880
- O75880
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together