Anti-CHRNA6 Rabbit Recombinant Polyclonal Antibody
Catalog # 77824-149
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Neuronal acetylcholine receptor subunit alpha-6 protein (26-239AA)
- Antigen Symbol:CHRNA6
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μl
- Form:Liquid
- Gene ID:Q15825
- Antigen Synonyms:Neuronal acetylcholine receptor subunit alpha-6
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: CHRNA6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human