Anti-OR2H1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: OR2H1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-294
- 77839-295
- Antigen Symbol:
- OR2H1
- OR2H1
- Antigen Name:
- Olfactory receptor 2H1 protein (157-193AA)
- Olfactory receptor 2H1 protein (157-193AA)
- Antigen Synonyms:
- OR2H1|OR2H6|OR2H8|Olfactory receptor 2H1|Hs6M1-16|OLFR42A-9004.14/9026.2|Olfactory receptor 2H6|Olfactory receptor 2H8|Olfactory receptor 6-2|OR6-2|Olfactory receptor OR6-32
- OR2H1|OR2H6|OR2H8|Olfactory receptor 2H1|Hs6M1-16|OLFR42A-9004.14/9026.2|Olfactory receptor 2H6|Olfactory receptor 2H8|Olfactory receptor 6-2|OR6-2|Olfactory receptor OR6-32
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZK4
- Q9GZK4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together