Anti-TBCB Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200, IP:1:200 to 1:2000
Type:
Antigen: TBCB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77806-541
- 77806-540
- Antigen Symbol:
- TBCB
- TBCB
- Antigen Name:
- Tubulin-folding cofactor B protein (1-244AA)
- Tubulin-folding cofactor B protein (1-244AA)
- Antigen Synonyms:
- CG22|CKAPI|Cytoskeleton associated protein 1|Cytoskeleton associated protein CKAPI|Cytoskeleton-associated protein 1|Cytoskeleton-associated protein CKAPI|Tbcb|TBCB_HUMAN|Tubulin folding cofactor B|Tubulin specific chaperone B|Tubulin-folding cofactor B|Tubulin-specific chaperone B
- CG22|CKAPI|Cytoskeleton associated protein 1|Cytoskeleton associated protein CKAPI|Cytoskeleton-associated protein 1|Cytoskeleton-associated protein CKAPI|Tbcb|TBCB_HUMAN|Tubulin folding cofactor B|Tubulin specific chaperone B|Tubulin-folding cofactor B|Tubulin-specific chaperone B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99426
- Q99426
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together