Anti-PTPN6 Rabbit Polyclonal Antibody
Catalog # 77807-265
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from SH-PTP1 around the phosphorylation site of Y536.
- Antigen Symbol:PTPN6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P29350
- Antigen Synonyms:70Z-SHP|EC 3.1.3.48|HCP|HCPH|Hematopoietic cell phosphatase|Hematopoietic cell protein tyrosine phosphatase|Hematopoietic cell protein-tyrosine phosphatase|HPTP1C|Protein tyrosine phosphatase 1C|Protein tyrosine phosphatase non receptor type 6|Protein tyrosine phosphatase SHP1|Protein-tyrosine phosphatase 1C|protein-tyrosine phosphatase SHP 1|Protein-tyrosine phosphatase SHP-1|PTN6_HUMAN|PTP 1C|PTP-1C|PTP1C|Ptpn6|SH PTP 1|SH PTP1|SH-PTP1|SHP 1|SHP 1L|SHP1|SHP1L|tyrosine protein phosphatase non receptor type 6|Tyrosine-protein phosphatase non-receptor type 6
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: PTPN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat