Anti-CYP11A1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: CYP11A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77807-938
- 77807-939
- Antigen Symbol:
- CYP11A1
- CYP11A1
- Antigen Name:
- Fusion protein of CYP11A1
- Fusion protein of CYP11A1
- Antigen Synonyms:
- Cholesterol 20 22 desmolase| Cholesterol desmolase| Cholesterol monooxygenase (side chain cleaving)| Cholesterol side chain cleavage enzyme| Cholesterol side chain cleavage enzyme mitochondrial| Cholesterol side-chain cleavage enzyme| CP11A_HUMAN| CYP11A| CYP11A1| CYPXIA1| Cytochrome P450 11A1| Cytochrome P450 11A1 mitochondrial| Cytochrome P450 family 11 subfamily A polypeptide 1| Cytochrome P450 subfamily XIA| Cytochrome P450(scc)| Cytochrome P450C11A1| mitochondrial| P450SCC| Steroid 20 22 lyase
- Cholesterol 20 22 desmolase| Cholesterol desmolase| Cholesterol monooxygenase (side chain cleaving)| Cholesterol side chain cleavage enzyme| Cholesterol side chain cleavage enzyme mitochondrial| Cholesterol side-chain cleavage enzyme| CP11A_HUMAN| CYP11A| CYP11A1| CYPXIA1| Cytochrome P450 11A1| Cytochrome P450 11A1 mitochondrial| Cytochrome P450 family 11 subfamily A polypeptide 1| Cytochrome P450 subfamily XIA| Cytochrome P450(scc)| Cytochrome P450C11A1| mitochondrial| P450SCC| Steroid 20 22 lyase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05108
- P05108
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together