Anti-PRSS23 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: PRSS23
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77795-650
- 77795-651
- Antigen Symbol:
- PRSS23
- PRSS23
- Antigen Name:
- Serine protease 23 protein (20-383AA)
- Serine protease 23 protein (20-383AA)
- Antigen Synonyms:
- Protease serine 23|PRS23_HUMAN|Prss23|Putative secreted protein ZSIG13|Serine protease 23|Serine protease umbilical endothelium|SIG13|SPUVE|UNQ270/PRO307|ZSIG13
- Protease serine 23|PRS23_HUMAN|Prss23|Putative secreted protein ZSIG13|Serine protease 23|Serine protease umbilical endothelium|SIG13|SPUVE|UNQ270/PRO307|ZSIG13
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95084
- O95084
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together