Anti-fbpB Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: fbpB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycobacterium kansasii
- Catalog No:
- 77815-489
- 77815-488
- Antigen Symbol:
- fbpB
- fbpB
- Antigen Name:
- Recombinant Mycobacterium kansasii Diacylglycerol acyltransferase/mycolyltransferase Ag85B protein (41-325AA)
- Recombinant Mycobacterium kansasii Diacylglycerol acyltransferase/mycolyltransferase Ag85B protein (41-325AA)
- Antigen Synonyms:
- fbpBDiacylglycerol acyltransferase/mycolyltransferase Ag85B|DGAT|EC 2.3.1.122|EC 2.3.1.20|30 kDa extracellular protein|Acyl-CoA:diacylglycerol acyltransferase|Antigen 85 complex B|85B|Ag85B|Extracellular alpha-antigen|Fibronectin-binding protein B|Fbps B
- fbpBDiacylglycerol acyltransferase/mycolyltransferase Ag85B|DGAT|EC 2.3.1.122|EC 2.3.1.20|30 kDa extracellular protein|Acyl-CoA:diacylglycerol acyltransferase|Antigen 85 complex B|85B|Ag85B|Extracellular alpha-antigen|Fibronectin-binding protein B|Fbps B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Mycobacterium kansasii
- Mycobacterium kansasii
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21160
- P21160
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together