Anti-RHBDF1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RHBDF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-983
- 77833-982
- Antigen Symbol:
- RHBDF1
- RHBDF1
- Antigen Name:
- Complement factor H-related protein 1 protein (132-221AA)
- Complement factor H-related protein 1 protein (132-221AA)
- Antigen Synonyms:
- RHBDF1|C16orf8|DIST1|IRHOM1|Inactive rhomboid protein 1|iRhom1|Epidermal growth factor receptor-related protein|Rhomboid 5 homolog 1|Rhomboid family member 1|p100hRho
- RHBDF1|C16orf8|DIST1|IRHOM1|Inactive rhomboid protein 1|iRhom1|Epidermal growth factor receptor-related protein|Rhomboid 5 homolog 1|Rhomboid family member 1|p100hRho
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96CC6
- Q96CC6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together