Anti-POLR3K Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: POLR3K
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77794-804
- 77794-803
- Antigen Symbol:
- POLR3K
- POLR3K
- Antigen Name:
- DNA-directed RNA polymerase III subunit RPC10 protein (1-108AA)
- DNA-directed RNA polymerase III subunit RPC10 protein (1-108AA)
- Antigen Synonyms:
- C11|C11 RNP3|DNA directed RNA polymerase III subunit K|DNA directed RNA polymerase III subunit RPC10|DNA directed RNA polymerases III 12.5 kDa polypeptide|DNA-directed RNA polymerase III subunit K|DNA-directed RNA polymerase III subunit RPC10|hRPC11|HsC11p|My010|POLR3K|Polymerase (RNA) III (DNA directed) polypeptide K|12.3 kDa|RNA polymerase III 12.5 kDa subunit|RNA polymerase III subunit (hRPC11)|RNA polymerase III subunit C10|RNA polymerase III subunit C11|RNA polymerase III subunit CII|RPC10|RPC10_HUMAN|RPC11|RPC12.5
- C11|C11 RNP3|DNA directed RNA polymerase III subunit K|DNA directed RNA polymerase III subunit RPC10|DNA directed RNA polymerases III 12.5 kDa polypeptide|DNA-directed RNA polymerase III subunit K|DNA-directed RNA polymerase III subunit RPC10|hRPC11|HsC11p|My010|POLR3K|Polymerase (RNA) III (DNA directed) polypeptide K|12.3 kDa|RNA polymerase III 12.5 kDa subunit|RNA polymerase III subunit (hRPC11)|RNA polymerase III subunit C10|RNA polymerase III subunit C11|RNA polymerase III subunit CII|RPC10|RPC10_HUMAN|RPC11|RPC12.5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2Y1
- Q9Y2Y1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together