Anti-SLC9A3R2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC9A3R2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-384
- 77793-385
- Antigen Symbol:
- SLC9A3R2
- SLC9A3R2
- Antigen Name:
- Na(+)/H(+) exchange regulatory cofactor NHE-RF2 protein (106-313AA)
- Na(+)/H(+) exchange regulatory cofactor NHE-RF2 protein (106-313AA)
- Antigen Synonyms:
- E3KARP|HGNC:11076|Isoform 3 regulator 2|Isoform 3 regulatory factor 2|MGC104639|Na(+)/H(+) exchange regulatory cofactor NHE RF2|Na(+)/H(+) exchange regulatory cofactor NHE-RF2|NHE3 kinase A regulatory protein|NHE3 kinase A regulatory protein E3KARP|NHE3 regulatory factor 2|NHE3RF2|NHERF 2|NHERF-2|NHERF2|NHRF2_HUMAN|OCTS2|SIP 1|SIP-1|SIP1|Slc9a3r2|Sodium hydrogen exchanger regulatory factor 2|Sodium-hydrogen exchanger regulatory factor 2|Sodium/hydrogen exchanger|Sodium/hydrogen exchanger regulatory factor 2|Solute carrier family 9 (sodium/hydrogen exchanger)|Solute carrier family 9 (sodium/hydrogen exchanger) isoform 3 regulator 2|Solute carrier family 9 (sodium/hydrogen exchanger) isoform 3 regulatory factor 2|Solute carrier family 9 (sodium/hydrogen exchanger) member 3 regulator 2|Solute carrier family 9 isoform 3 regulator 2|Solute carrier family 9 isoform A3 regulatory factor 2|SRY interacting protein 1|SRY-interacting protein 1|TKA 1|TKA-1|TKA1|Tyrosine kinase activator protein 1
- E3KARP|HGNC:11076|Isoform 3 regulator 2|Isoform 3 regulatory factor 2|MGC104639|Na(+)/H(+) exchange regulatory cofactor NHE RF2|Na(+)/H(+) exchange regulatory cofactor NHE-RF2|NHE3 kinase A regulatory protein|NHE3 kinase A regulatory protein E3KARP|NHE3 regulatory factor 2|NHE3RF2|NHERF 2|NHERF-2|NHERF2|NHRF2_HUMAN|OCTS2|SIP 1|SIP-1|SIP1|Slc9a3r2|Sodium hydrogen exchanger regulatory factor 2|Sodium-hydrogen exchanger regulatory factor 2|Sodium/hydrogen exchanger|Sodium/hydrogen exchanger regulatory factor 2|Solute carrier family 9 (sodium/hydrogen exchanger)|Solute carrier family 9 (sodium/hydrogen exchanger) isoform 3 regulator 2|Solute carrier family 9 (sodium/hydrogen exchanger) isoform 3 regulatory factor 2|Solute carrier family 9 (sodium/hydrogen exchanger) member 3 regulator 2|Solute carrier family 9 isoform 3 regulator 2|Solute carrier family 9 isoform A3 regulatory factor 2|SRY interacting protein 1|SRY-interacting protein 1|TKA 1|TKA-1|TKA1|Tyrosine kinase activator protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15599
- Q15599
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together