Anti-GTF2H2 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GTF2H2
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-521
- 77823-522
- Antigen Symbol:
- GTF2H2
- GTF2H2
- Antigen Name:
- General transcription factor IIH subunit 2 protein (1-157AA)
- General transcription factor IIH subunit 2 protein (1-157AA)
- Antigen Synonyms:
- Basic transcription factor 2 44 kDa subunit|BTF2|BTF2 p44|BTF2-p44|BTF2P44|General transcription factor IIH|General transcription factor IIH polypeptide 2|General transcription factor IIH subunit 2|polypeptide 2|44kDa|Gtf2h2|MGC102806|p44|T BTF2P44|T-BTF2P44|TF2H2_HUMAN|TFIIH|TFIIH basal transcription factor complex p44 subunit
- Basic transcription factor 2 44 kDa subunit|BTF2|BTF2 p44|BTF2-p44|BTF2P44|General transcription factor IIH|General transcription factor IIH polypeptide 2|General transcription factor IIH subunit 2|polypeptide 2|44kDa|Gtf2h2|MGC102806|p44|T BTF2P44|T-BTF2P44|TF2H2_HUMAN|TFIIH|TFIIH basal transcription factor complex p44 subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13888
- Q13888
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together