Anti-MICU1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MICU1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-084
- 77835-085
- Antigen Symbol:
- MICU1
- MICU1
- Antigen Name:
- Calcium uptake protein 1, mitochondrial protein (55-191AA)
- Calcium uptake protein 1, mitochondrial protein (55-191AA)
- Antigen Synonyms:
- Allergen Hom s 4|ara CALC|atopy related autoantigen|atopy related autoantigen CALC|Atopy-related autoantigen CALC|CALC|calcium binding atopy related autoantigen 1|Calcium uptake protein 1|calcium uptake protein 1, mitochondrial|Calcium-binding atopy-related autoantigen 1|CBARA1|EFHA3|FLJ12684|MICU1|MICU1_HUMAN|mitochondrial|mitochondrial calcium uptake 1|MPXPS
- Allergen Hom s 4|ara CALC|atopy related autoantigen|atopy related autoantigen CALC|Atopy-related autoantigen CALC|CALC|calcium binding atopy related autoantigen 1|Calcium uptake protein 1|calcium uptake protein 1, mitochondrial|Calcium-binding atopy-related autoantigen 1|CBARA1|EFHA3|FLJ12684|MICU1|MICU1_HUMAN|mitochondrial|mitochondrial calcium uptake 1|MPXPS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BPX6
- Q9BPX6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together