Anti-ALYREF Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ALYREF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-706
- 77832-707
- Antigen Symbol:
- ALYREF
- ALYREF
- Antigen Name:
- THO complex subunit 4 protein (186-240AA)
- THO complex subunit 4 protein (186-240AA)
- Antigen Synonyms:
- Ally of AML-1 and LEF-1|Ally of AML1 and LEF1|ALY|ALY/REF|Aly/REF export factor|BEF|bZIP enhancing factor|bZIP-enhancing factor BEF|REF|THO complex 4|THO complex subunit 4|Tho4|thoc4|THOC4_HUMAN|Transcriptional coactivator Aly/REF|Transcriptional coactivator
- Ally of AML-1 and LEF-1|Ally of AML1 and LEF1|ALY|ALY/REF|Aly/REF export factor|BEF|bZIP enhancing factor|bZIP-enhancing factor BEF|REF|THO complex 4|THO complex subunit 4|Tho4|thoc4|THOC4_HUMAN|Transcriptional coactivator Aly/REF|Transcriptional coactivator
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86V81
- Q86V81
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together