Anti-DDB1 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DDB1
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-253
- 77823-252
- Antigen Symbol:
- DDB1
- DDB1
- Antigen Name:
- DNA damage-binding protein 1 protein (741-943AA)
- DNA damage-binding protein 1 protein (741-943AA)
- Antigen Synonyms:
- Damage specific DNA binding protein 1|Damage-specific DNA-binding protein 1|DDB 1|DDB p127 subunit|Ddb1|DDB1_HUMAN|DDBa|DNA damage binding protein 1|DNA damage-binding protein 1|DNA damage-binding protein a|HBV X-associated protein 1|UV damaged DNA binding factor|UV damaged DNA binding protein 1|UV DDB 1|UV DDB1|UV-damaged DNA-binding factor|UV-damaged DNA-binding protein 1|UV-DDB 1|X associated protein 1|XAP 1|XAP-1|XAP1|Xeroderma pigmentosum group E complementing protein|Xeroderma pigmentosum group E-complementing protein|XPCe|XPE|XPE BF|XPE binding factor|XPE-BF|XPE-binding factor
- Damage specific DNA binding protein 1|Damage-specific DNA-binding protein 1|DDB 1|DDB p127 subunit|Ddb1|DDB1_HUMAN|DDBa|DNA damage binding protein 1|DNA damage-binding protein 1|DNA damage-binding protein a|HBV X-associated protein 1|UV damaged DNA binding factor|UV damaged DNA binding protein 1|UV DDB 1|UV DDB1|UV-damaged DNA-binding factor|UV-damaged DNA-binding protein 1|UV-DDB 1|X associated protein 1|XAP 1|XAP-1|XAP1|Xeroderma pigmentosum group E complementing protein|Xeroderma pigmentosum group E-complementing protein|XPCe|XPE|XPE BF|XPE binding factor|XPE-BF|XPE-binding factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16531
- Q16531
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together