Order Entry
United States
ContactUsLinkComponent
 
Anti-GPR17 Rabbit Recombinant Polyclonal Antibody
New Product
 
undefined
Anti-GPR17 Rabbit Recombinant Polyclonal Antibody
New Product

 

  • Catalog No:
  • 77823-875
  • 77823-876
  • Antigen Symbol:
  • GPR17
  • GPR17
  • Antigen Name:
  • Uracil nucleotide/cysteinyl leukotriene receptor protein (1-36AA)
  • Uracil nucleotide/cysteinyl leukotriene receptor protein (1-36AA)
  • Antigen Synonyms:
  • DKFZp686M18273|G protein coupled receptor 17|G-protein coupled receptor 17|GPR 17|Gpr17|GPR17_HUMAN|P2Y like receptor|P2Y-like receptor|Probable P2Y purinoceptor GPR 17|Probable P2Y purinoceptor GPR17|R12|UDP/CysLT receptor|Uracil nucleotide/cysteinyl leukotriene receptor
  • DKFZp686M18273|G protein coupled receptor 17|G-protein coupled receptor 17|GPR 17|Gpr17|GPR17_HUMAN|P2Y like receptor|P2Y-like receptor|Probable P2Y purinoceptor GPR 17|Probable P2Y purinoceptor GPR17|R12|UDP/CysLT receptor|Uracil nucleotide/cysteinyl leukotriene receptor
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q13304
  • Q13304
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together