Anti-USH1G Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: USH1G
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77826-572
- 77826-573
- Antigen Symbol:
- USH1G
- USH1G
- Antigen Name:
- Usher syndrome type-1G protein (1-300AA)
- Usher syndrome type-1G protein (1-300AA)
- Antigen Synonyms:
- ANKS 4A|ANKS4A|FLJ33924|SANS|Scaffold protein containing ankyrin repeats and SAM domain|USH 1G|USH1G_HUMAN|Usher syndrome 1G (autosomal recessive)|Usher syndrome type 1G protein|Usher syndrome type-1G protein
- ANKS 4A|ANKS4A|FLJ33924|SANS|Scaffold protein containing ankyrin repeats and SAM domain|USH 1G|USH1G_HUMAN|Usher syndrome 1G (autosomal recessive)|Usher syndrome type 1G protein|Usher syndrome type-1G protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q495M9
- Q495M9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together